2-(4-Bromo-2,6-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

31086-25G 31086-5G
76887-078EA 524.09 USD
76887-078 76887-080
2-(4-Bromo-2,6-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Bromo-2,6-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₂H₁₄BBrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD26143393
CAS Number: 1799485-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR