2,4-Dibromo-5-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

30837-1G 30837-25G 30837-500MG 30837-5G
76886-994EA 290.69 USD
76886-994 76886-996 76886-998 76887-000
2,4-Dibromo-5-fluorobenzaldehyde ≥95%
2,4-Dibromo-5-fluorobenzaldehyde
Formula: C₇H₃Br₂FO
Storage Temperature: Refrigerator
MDL Number: MFCD28784588
CAS Number: 1806352-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR