1-Iodo-3,5-dimethoxybenzene ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Dimethoxy-5-iodobenzene

30802-25G 30802-5G
76886-982EA 399.96 USD
76886-982 76886-984
1-Iodo-3,5-dimethoxybenzene ≥95% research grade
1-Iodo-3,5-dimethoxybenzene
Formula: C₈H₉IO₂
MW: 264.0603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08729270
CAS Number: 25245-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR