2,4-Dichloro-3-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

30785-1G 30785-25G 30785-5G
76886-972EA 62.6 USD
76886-972 76886-974 76886-976
2,4-Dichloro-3-fluoropyridine ≥97%
2,4-Dichloro-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂FN
Storage Temperature: Refrigerator
MDL Number: MFCD12196994
CAS Number: 628691-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR