1-Chloro-5-fluoro-2-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

30684-1G 30684-25G 30684-500MG 30684-5G
76886-958EA 185.66 USD
76886-958 76886-960 76886-962 76886-964
1-Chloro-5-fluoro-2-iodo-3-methylbenzene ≥97%
1-Chloro-5-fluoro-2-iodo-3-methylbenzene
Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16817667
CAS Number: 1242339-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR