(5-(tert-Butyl)-2-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

30635-1G 30635-500MG 30635-5G
76886-950EA 563.34 USD
76886-950 76886-952 76886-954
(5-(tert-Butyl)-2-methylphenyl)boronic acid ≥97% research grade
(5-(tert-Butyl)-2-methylphenyl)boronic acid
Formula: C₁₁H₁₇BO₂
MW: 192.062 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32065678
CAS Number: 2377606-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR