2-(4-Methoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(2-Pyridyl)anisole

30416-25G 30416-5G
76886-900EA 230.22 USD
76886-900 76886-902
2-(4-Methoxyphenyl)pyridine ≥97%
2-(4-Methoxyphenyl)pyridine
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092333
CAS Number: 5957-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR