1,3-Dibromo-2-chlorobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

30316-25G 30316-5G
76886-892EA 190.97 USD
76886-892 76886-894
1,3-Dibromo-2-chlorobenzene ≥97% research grade
1,3-Dibromo-2-chlorobenzene
Formula: C₆H₃Br₂Cl
Storage Temperature: Ambient
MDL Number: MFCD12922600
CAS Number: 19230-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR