3-Bromo-2,6-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30277-1G 30277-25G 30277-5G
76886-882EA 117.76 USD
76886-882 76886-884 76886-886
3-Bromo-2,6-difluorobenzyl alcohol ≥97%
3-Bromo-2,6-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD20483917
CAS Number: 438050-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR