5,6-Dichloropyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5,6-Dichloro-3-pyridinol, 2,3-Dichloro-5-hydroxypyridine

29786-1G 29786-25G 29786-5G
76886-820EA 73.21 USD
76886-820 76886-822 76886-824
5,6-Dichloropyridin-3-ol ≥97%
5,6-Dichloropyridin-3-ol
Formula: C₅H₃Cl₂NO
MW: 163.9900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835219
CAS Number: 110860-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR