2-Fluoro-5-methylanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-methoxytoluene, 1-Fluoro-2-methoxy-4-methylbenzene

29726-25G 29726-5G
76886-810EA 183.54 USD
76886-810 76886-812
2-Fluoro-5-methylanisole ≥97%
2-Fluoro-5-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665791
CAS Number: 63762-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR