2,3-Difluoro-6-iodo-1-(trimethylsilyl)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

29661-1G 29661-25G 29661-5G
76886-796EA 62.6 USD
76886-796 76886-798 76886-800
2,3-Difluoro-6-iodo-1-(trimethylsilyl)benzene ≥97% research grade
2,3-Difluoro-6-iodo-1-(trimethylsilyl)benzene
Formula: C₉H₁₁F₂ISi
Storage Temperature: Ambient
MDL Number: MFCD28977323
CAS Number: 1807646-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR