1-(2-Bromopyridin-4-yl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
29647-1G
29647-25G
29647-500MG
29647-5G
76886-780EA
185.66
USD
76886-780
76886-782
76886-784
76886-786
1-(2-Bromopyridin-4-yl)ethanone ≥97%
1-(2-Bromopyridin-4-yl)ethanone
Precaución: for Research Use Only
Formula:
C₇H₆BrNO MW: 200.033 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07367876 CAS Number: 864674-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...