2',3'-dichloro-[1,1'-biphenyl]-4-ol ≥97%

Supplier: AOBChem USA

Warning

29549-1G 29549-25G 29549-500MG 29549-5G
76886-750EA 183.54 USD
76886-750 76886-752 76886-754 76886-756
2',3'-dichloro-[1,1'-biphenyl]-4-ol ≥97%
2',3'-dichloro-[1,1'-biphenyl]-4-ol
Formula: C₁₂H₈Cl₂O
MW: 239.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11932358
CAS Number: 53714-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR