4-Methoxy-2-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Methoxy-2-(trifluoromethyl)phenyl)methanol

29209-1G 29209-25G 29209-5G
76886-692EA 62.6 USD
76886-692 76886-694 76886-696
4-Methoxy-2-(trifluoromethyl)benzyl alcohol ≥97%
4-Methoxy-2-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06202983
CAS Number: 773871-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR