[2-(Benzyloxy)-4-methoxyphenyl]methanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-(Benzyloxy)-4-methoxyphenyl)methanol

29187-1G 29187-25G 29187-500MG 29187-5G
76886-676EA 165.51 USD
76886-676 76886-678 76886-680 76886-682
[2-(Benzyloxy)-4-methoxyphenyl]methanol ≥95%
[2-(Benzyloxy)-4-methoxyphenyl]methanol
Formula: C₁₅H₁₆O₃
MW: 244.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922687
CAS Number: 171817-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR