(4-(difluoromethoxy)-2-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Difluoromethoxy)-2-fluorophenyl)boronic acid

29171-1G 29171-500MG 29171-5G 29171-25G
76886-650EA 519.85 USD
76886-650 76886-652 76886-654 77658-504
(4-(difluoromethoxy)-2-fluorophenyl)boronic acid ≥97%
(4-(difluoromethoxy)-2-fluorophenyl)boronic acid
Formula: C₇H₆BF₃O₃
MW: 205.927 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18395053
CAS Number: 1004775-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR