2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-(Cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester
29118-1G
29118-25G
29118-500MG
29118-5G
76886-618EA
124.13
USD
76886-618
76886-620
76886-622
76886-624
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester ≥97%
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₂BNO₃ MW: 275.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07781184 CAS Number: 947191-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...