1-(4-Bromo-3-methoxyphenyl)-1H-imidazole ≥97%

Supplier: AOBChem USA

Warning

28740-25G 28740-5G
76886-552EA 346.92 USD
76886-552 76886-554
1-(4-Bromo-3-methoxyphenyl)-1H-imidazole ≥97%
1-(4-Bromo-3-methoxyphenyl)-1H-imidazole
Formula: C₁₀H₉BrN₂O
MW: 253.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28360704
CAS Number: 1562339-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR