1-(Methoxymethoxy)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

28719-1G 28719-25G 28719-500MG 28719-5G
76886-536EA 152.77 USD
76886-536 76886-538 76886-540 76886-542
1-(Methoxymethoxy)-2-methylbenzene ≥97%
1-(Methoxymethoxy)-2-methylbenzene
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00962976
CAS Number: 55359-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR