1-Bromo-2-cyclopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

28656-1G 28656-25G 28656-500MG 28656-5G
76886-510EA 120.95 USD
76886-510 76886-512 76886-514 76886-516
1-Bromo-2-cyclopropylbenzene ≥97%
1-Bromo-2-cyclopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₉Br
Storage Temperature: Ambient
MDL Number: MFCD11109720
CAS Number: 57807-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR