(4-Cyclobutylphenyl)boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

28587-1G 28587-25G 28587-500MG 28587-5G
76886-494EA 274.78 USD
76886-494 76886-496 76886-498 76886-500
(4-Cyclobutylphenyl)boronic acid pinacol ester ≥97% research grade
(4-Cyclobutylphenyl)boronic acid pinacol ester
Formula: C₁₆H₂₃BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18733634
CAS Number: 2121513-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR