1-(4-Bromophenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

28513-1G 28513-25G 28513-500MG 28513-5G
76886-462EA 129.43 USD
76886-462 76886-464 76886-466 76886-468
1-(4-Bromophenyl)cyclobutanol ≥97%
1-(4-Bromophenyl)cyclobutanol
Formula: C₁₀H₁₁BrO
MW: 227.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18205682
CAS Number: 19936-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR