Methyl 2,4,6-trichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4,6-Trichlorobenzoic acid methyl ester

28443-1G 28443-25G 28443-500MG 28443-5G
76886-420EA 185.66 USD
76886-420 76886-422 76886-424 76886-426
Methyl 2,4,6-trichlorobenzoate ≥97%
Methyl 2,4,6-trichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Cl₃O₂
MW: 239.483 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12024963
CAS Number: 86569-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR