2,4,6-Trichlorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

28368-25G 28368-5G
76886-414EA 283.27 USD
76886-414 76886-416
2,4,6-Trichlorobenzyl alcohol ≥97%
2,4,6-Trichlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅Cl₃O
MW: 211.47
Storage Temperature: Ambient
MDL Number: MFCD03093891
CAS Number: 217479-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR