2-(2,5-bis(Trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

28192-1G 28192-25G 28192-5G 28192-500MG
76886-392EA 112.46 USD
76886-392 76886-394 76886-396 77649-774
2-(2,5-bis(Trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,5-bis(Trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₄H₁₅BF₆O₂
MW: 340.069 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12405523
CAS Number: 1073339-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR