(4-Fluorobenzofuran-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

28182-1G 28182-500MG 28182-5G
76886-382EA 973.91 USD
76886-382 76886-384 76886-386
(4-Fluorobenzofuran-2-yl)boronic acid ≥97%
(4-Fluorobenzofuran-2-yl)boronic acid
Formula: C₈H₆BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28154569
CAS Number: 1423791-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR