1-BOC-4-Bromo-3-methylpyrazole ≥97%

Supplier: AOBChem USA

Warning

28151-25G 28151-5G 28151-1G
76886-374EA 453.01 USD
76886-374 76886-376 77641-706
1-BOC-4-Bromo-3-methylpyrazole ≥97%
1-BOC-4-Bromo-3-methylpyrazole
Formula: C₉H₁₃BrN₂O₂
MW: 261.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18434446
CAS Number: 1021919-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR