1-(7-Nitro-3,4-dihydroquinolin-1(2H)-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

28089-1G 28089-25G 28089-500MG 28089-5G
76886-346EA 203.7 USD
76886-346 76886-348 76886-350 76886-352
1-(7-Nitro-3,4-dihydroquinolin-1(2H)-yl)ethanone ≥97%
1-(7-Nitro-3,4-dihydroquinolin-1(2H)-yl)ethanone
Formula: C₁₁H₁₂N₂O₃
MW: 220.225 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00496671
CAS Number: 40484-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR