1,5-Difluoro-2-iodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

27955-1G 27955-25G 27955-5G
76886-304EA 84.88 USD
76886-304 76886-306 76886-308
1,5-Difluoro-2-iodo-4-methylbenzene ≥97%
1,5-Difluoro-2-iodo-4-methylbenzene
Formula: C₇H₅F₂I
MW: 254.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658718
CAS Number: 333447-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR