2-(3,4-Difluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

27744-1G 27744-25G 27744-5G
76886-260EA 62.6 USD
76886-260 76886-262 76886-264
2-(3,4-Difluorophenyl)-1,3-dioxolane ≥97%
2-(3,4-Difluorophenyl)-1,3-dioxolane
Formula: C₉H₈F₅O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06209066
CAS Number: 773101-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR