1-Bromo-4-iodo-2-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-iodo-α,α,α-trifluoroanisole, 2-Bromo-5-iodophenyl trifluoromethyl ether

27705-1G 27705-25G 27705-500MG 27705-5G
76886-248EA 203.7 USD
76886-248 76886-250 76886-252 76886-254
1-Bromo-4-iodo-2-(trifluoromethoxy)benzene ≥97%
1-Bromo-4-iodo-2-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₃IO
MW: 366.902 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12761590
CAS Number: 1187984-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR