(5-Methoxy-2-methylphenyl)methanol 97%

Supplier: AOBChem USA

Warning

27594-1G 27594-25G 27594-5G 27594-500MG
76886-234EA 94.43 USD
76886-234 76886-236 76886-238 77652-568
(5-Methoxy-2-methylphenyl)methanol 97%
(5-Methoxy-2-methylphenyl)methanol
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18398313
CAS Number: 73502-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR