2-Bromo-4,6-dichloroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-3,5-dichloro-2-iodobenzene

27540-1G 27540-25G 27540-5G
76886-220EA 65.78 USD
76886-220 76886-222 76886-224
2-Bromo-4,6-dichloroiodobenzene ≥97%
2-Bromo-4,6-dichloroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂I
Storage Temperature: Refrigerator
CAS Number: 81067-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR