3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

27245-1G 27245-25G 27245-500MG 27245-5G
76886-180EA 144.29 USD
76886-180 76886-182 76886-184 76886-186
3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester ≥97% research grade
3-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₈BFO₃
Storage Temperature: Ambient
MDL Number: MFCD18732617
CAS Number: 1082066-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR