(5-Fluoro-4-methylpyridin-3-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

27141-1G 27141-500MG 27141-5G
76886-148EA 765.97 USD
76886-148 76886-150 76886-152
(5-Fluoro-4-methylpyridin-3-yl)boronic acid ≥95%
(5-Fluoro-4-methylpyridin-3-yl)boronic acid
Formula: C₆H₇BFNO₂
MW: 154.935 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18258205
CAS Number: 1416500-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR