2,4-Dibromo-3,6-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

26836-25G 26836-5G
76886-094EA 360.71 USD
76886-094 76886-096
2,4-Dibromo-3,6-dimethylaniline ≥97%
2,4-Dibromo-3,6-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₉Br₂N
Storage Temperature: Ambient
MDL Number: MFCD08689499
CAS Number: 26829-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR