1,2-Dimethylbenzimidazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,2-Dimethyl-1H-benzo[d]imidazole

26820-1G 26820-25G 26820-5G
76886-080EA 64.72 USD
76886-080 76886-082 76886-084
1,2-Dimethylbenzimidazole ≥97%
1,2-Dimethylbenzimidazole
Formula: C₉H₁₀N₂
MW: 146.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022840
CAS Number: 2876-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR