1-Bromo-3-chloro-4-methyl-2-(methylthio)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

26728-25G 26728-5G
76886-068EA 418 USD
76886-068 76886-070
1-Bromo-3-chloro-4-methyl-2-(methylthio)benzene ≥97% research grade
1-Bromo-3-chloro-4-methyl-2-(methylthio)benzene
Formula: C₈H₈BrClS
MW: 251.571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756673
CAS Number: 1809168-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR