1-(3,4-Dichloro-benzyl)-piperidine ≥97%

Supplier: AOBChem USA

Warning

26584-1G 26584-25G 26584-500MG 26584-5G
76886-026EA 231.28 USD
76886-026 76886-028 76886-030 76886-032
1-(3,4-Dichloro-benzyl)-piperidine ≥97%
1-(3,4-Dichloro-benzyl)-piperidine
Formula: C₁₂H₁₅Cl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD06080743
CAS Number: 891401-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR