(2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
26535-1G
26535-25G
26535-500MG
26535-5G
76886-014EA
124.13
USD
76886-014
76886-016
76886-018
76886-020
(2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol ≥97%
(2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol
Formula:
C₁₃H₁₈BFO₃ MW: 252.08 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18729954 CAS Number: 1352733-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...