(5-Methylpyridin-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

26485-1G 26485-25G 26485-5G
76886-000EA 84.88 USD
76886-000 76886-002 76886-004
(5-Methylpyridin-2-yl)methanol ≥97%
(5-Methylpyridin-2-yl)methanol
Formula: C₇H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD09999992
CAS Number: 22940-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR