(2-Fluoro-5-methoxypyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

26278-1G 26278-500MG 26278-5G
76885-958EA 558.04 USD
76885-958 76885-960 76885-962
(2-Fluoro-5-methoxypyridin-3-yl)methanol ≥97%
(2-Fluoro-5-methoxypyridin-3-yl)methanol
Formula: C₇H₈FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16611250
CAS Number: 1227511-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR