(3-Bromo-5-fluoropyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

26257-1G 26257-25G 26257-500MG 26257-5G
76885-944EA 218.55 USD
76885-944 76885-946 76885-948 76885-950
(3-Bromo-5-fluoropyridin-4-yl)methanol ≥97%
(3-Bromo-5-fluoropyridin-4-yl)methanol
Formula: C₆H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD16607001
CAS Number: 1227586-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR