(4-(Benzyloxy)-3-fluorophenyl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

26168-1G 26168-25G 26168-500MG 26168-5G
76885-928EA 124.13 USD
76885-928 76885-930 76885-932 76885-934
(4-(Benzyloxy)-3-fluorophenyl)methanol ≥97% research grade
(4-(Benzyloxy)-3-fluorophenyl)methanol
Formula: C₁₄H₁₃FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11190327
CAS Number: 536974-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR