1-Bromo-4-(methoxymethyl)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

26055-1G 26055-25G 26055-500MG 26055-5G
76885-906EA 231.28 USD
76885-906 76885-908 76885-910 76885-912
1-Bromo-4-(methoxymethyl)-2-methylbenzene ≥97%
1-Bromo-4-(methoxymethyl)-2-methylbenzene
Formula: C₉H₁₁BrO
Storage Temperature: Refrigerator
MDL Number: MFCD24848698
CAS Number: 166386-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR