1H-Pyrazolo[3,4-b]pyridin-4-ylhydrazine ≥97%

Supplier: AOBChem USA

Warning

26013-1G 26013-25G 26013-500MG 26013-5G
76885-866EA 379.81 USD
76885-866 76885-868 76885-870 76885-872
1H-Pyrazolo[3,4-b]pyridin-4-ylhydrazine ≥97%
1H-Pyrazolo[3,4-b]pyridin-4-ylhydrazine
Formula: C₆H₇N₅
MW: 149.153 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13178151
CAS Number: 49834-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR