[3-Fluoro-4-(Methylsulfanyl)Phenyl]Methanol ≥97%

Supplier: AOBChem USA

Warning

25963-1G 25963-25G 25963-500MG 25963-5G
76885-836EA 201.58 USD
76885-836 76885-838 76885-840 76885-842
[3-Fluoro-4-(Methylsulfanyl)Phenyl]Methanol ≥97%
[3-Fluoro-4-(Methylsulfanyl)Phenyl]Methanol
Formula: C₈H₉FOS
Storage Temperature: Ambient
MDL Number: MFCD18891415
CAS Number: 252562-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR