2-Bromo-4-chloro-6-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-4-chloro-6-fluorophenyl)methanol

25939-1G 25939-500MG 25939-5G 25939-25G
76885-806EA 498.63 USD
76885-806 76885-808 76885-810 77654-506
2-Bromo-4-chloro-6-fluorobenzyl alcohol ≥97%
2-Bromo-4-chloro-6-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24848700
CAS Number: 1449008-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR