4-Bromo-3-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

25667-25G 25667-5G
76885-752EA 247.19 USD
76885-752 76885-754
4-Bromo-3-fluorobenzyl alcohol ≥97%
4-Bromo-3-fluorobenzyl alcohol
Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD08236860
CAS Number: 222978-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR